Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container, BioTitan retainers, three ports, Luer locks
Shop All Thermo Scientific Products

Description
The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.
Features of the Thermo Scientific 2D BPC system include:
- Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
- Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
- Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Thermo Scientific 2D BPC include:
- Harvest from, feed into, and collect samples from bioreactors and fermentors
- Buffer, culture media, and process liquid preparation and storage
- Chromatography feed and fraction collection
- Harvest, storage, and transport of bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library with over 20,000 unique components that include:
- Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
- Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
- Filters—used for bioburden reduction; sterilization grade filters and vent filters
- Tubing—thermoplastic elastomers, platinum-cured silicone
- Sample ports—septum-style and needle-free ports
- Clamps—sliders, pinch, and crimp bands
Specifications
Specifications
| Green Features | ISCC PLUS |
| Capacity (Metric) | 10 L |
| Quantity | 10 L |
| Tubing Style | TPE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.