Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), three ports, MPCs, quick-connect
GREENER_CHOICE

Catalog No. SH3100505 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
2 L
5 L
10 L
20 L
50 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3100505 2 L
SH3100501 5 L
SH3100502 10 L
SH3100503 20 L
SH3100504 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SH3100505 Supplier Thermo Scientific™ Supplier No. SH3100505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis5-14 single-web, multi-layer film.

Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ISO 7
Color Transparent
Disposable Single-use
For Use With (Application) Bioprocessing
Sterility Sterile
Tubing Style C-Flex
Film Description Aegis 5-14
Port Detail 3 ports
Quantity 2 L
Capacity (Metric) 2 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.