Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 250mL with 2 Ports, Luer Lock, 2D BPC
GREENER_CHOICE

Catalog No. p-7184450
Change view
Click to view available options
Capacity (Metric):
250 mL
Tubing Style:
12 in. of 1/4 in. Tubing
No Tubing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Tubing Style
SH3065713 250 mL No Tubing
SH3065813 250 mL 12 in. of 1/4 in. Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3065713 Supplier Thermo Scientific™ Supplier No. SH30657.13
Only null left
Add to Cart
Add to Cart

Simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Using Thermo Scientific™ Labtainer™ BPC bag with 2 top ports (luer lock and MPC insert), simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance, and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation systems

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport &

    Shipping - bulk sterile liquids, buffers, media and process intermediates within and between facilities

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Dimensions 19.1 cm x 15 cm (7.5 in. x 5.9 in.)
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity 10 per case
Sterilization Method Gamma Irradiated
Tubing Line 1 Line 1. Body luer lock connection, polypropylene. No Tubing
Tubing Line 2 Line 2. Insert luer lock connection, polypropylene. No Tubing Line
Product Line Labtainer
Port Detail Edge Ports: 2 x 0.25 in.
Film Description CX5-14
Tubing Style No Tubing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.