Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors
GREENER_CHOICE

Catalog No. SS00103I
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00103I 50 L
SS00104I 100 L
SS00105I 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SS00103I Supplier Thermo Scientific™ Supplier No. SS00103I
Add to Cart
Add to Cart

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 MPC insert + cap, 1 MPC body + cap). It is available in 50, 100 and 200L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: MPC insert with 9.5mm (3/8 in.) barb, cap, and pinch clamp. Tubing: Silicone, length: 51cm (20 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
  • Line 2: MPC body with 9.5mm (3/8 in.) barb, plug, and pinch clamp.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) TK-0500-05
Includes 2 Ports (1 MPC Insert + Cap, 1 MPC Body + Cap)
Material Polyethylene
Tubing Style C-Flex, 51 cm (20 in.)
Film Description ASI™ 26/77
Outer Packaging 4 mil. poly bag, One 4-mil. poly bag
Port Detail MPC connections + caps, pinch clamps
Product Line Labtainer
Protective Inner Packaging Poly bags on tubing ends, protective material around rough areas
Quantity 10/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 95.3 x 54.6 cm (37.5 x 21.5 in.)
Length (English) 37.5 in.
Width (English) 21.5 in.
Product Type Bioprocess Container
Capacity (Metric) 50 L
Length (Metric) 95.3 cm
Width (Metric) 54.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.