Learn More
Labtech Inc LabTech EV400H manual rotary evaporator, Speed Range: 20 - 260 rmp, Temperature range: Ambient - 180℃, Temp. Control Mode: PID, Heating power: 1300W, Bath material: PTFE coating/SS, Power Supply: 115V, 60Hz
Catalog No. 5000496186
Encompass_Preferred
LabTech EV400H manual rotary evaporator, Speed Range: 20 - 260 rmp, Temperature range: Ambient - 180℃, Temp. Control Mode: PID, Heating power: 1300W, Bath material: PTFE coating/SS, Power Supply: 115V, 60Hz
Catalog No. 50-004-96186 Supplier Labtech Inc Supplier No. LEV400H
LabTech EV400H manual rotary evaporator, Speed Range: 20 - 260 rmp, Temperature range: Ambient - 180℃, Temp. Control Mode: PID, Heating power: 1300W, Bath material: PTFE coating/SS, Power Supply: 115V, 60Hz