Learn More
Electron Microscopy Sciences Lab-Tek II Coverglass, 1 Chamber, 12 Trays, Sterile
SDP
Catalog No. 501899267
Encompass_Preferred
Intended for high magnification examination of living cells using a microscope. The thin glass growth surface allows optimal optical clarity when viewing living cells with high resolution of an inverted microscope. The Chambered Coverglass is provided sterile and ready to use. Media chambers cannot be removed from the coverglass surface.
Type: 1 Chamber
Package: 12 Trays
Sterile: Sterile
Catalog No. 50-189-9267 Supplier Electron Microscopy Sciences Supplier No. 7037812
Intended for high magnification examination of living cells using a microscope. The thin glass growth surface allows optimal optical clarity when viewing living cells with high resolution of an inverted microscope. The Chambered Coverglass is provided sterile and ready to use. Media chambers cannot be removed from the coverglass surface.
Type: 1 Chamber
Package: 12 Trays
Sterile: Sterile