Learn More
Electron Microscopy Sciences Lab-Tek 1 Chamber Slide, Glass, 6 trays
SDP
Catalog No. 501899288
Encompass_Preferred
Chamber slides combine a media-containing chambered super-structure with a standard soda lime glass slide or Permanox plastic slide, sealing the two together with a biologically inert silicone gasket. After cells are grown, the chamber and gasket are easily removed. The slide may then be fixed, stained, cover slipped, and filed with the usual convenience. These slides, once fixed and stained, can be embedded.
Packed in a sterile sealed tray, 8 units per tray. Glass does not interfere with fluorescent label.
Material: Glass
Package: 6 trays
Catalog No. 50-189-9288 Supplier Electron Microscopy Sciences Supplier No. 70384
Chamber slides combine a media-containing chambered super-structure with a standard soda lime glass slide or Permanox plastic slide, sealing the two together with a biologically inert silicone gasket. After cells are grown, the chamber and gasket are easily removed. The slide may then be fixed, stained, cover slipped, and filed with the usual convenience. These slides, once fixed and stained, can be embedded.
Packed in a sterile sealed tray, 8 units per tray. Glass does not interfere with fluorescent label.
Material: Glass
Package: 6 trays