Learn More
Thomas Scientific Bungee Cord Pack 7" To 18" Labungee
SDP
Catalog No. 501461129
Encompass
The BenchRocker is designed to provide precise speeds and tilt angles for a broad range of molecular and biological mixing applications. The user has the ability to adjust parameters to match the vessel size as well as the volume of liquid being mixed, yielding optimum results. The rocking speed and tilt angle are easily adjustable. At low speeds and minimal tilt angles, the BenchRocker 2D provides a gentle wave effect for non-foaming mixing for blot washing and gel work.
Catalog No. 50-146-1129 Supplier Thomas Scientific Supplier No. 1227U69
The BenchRocker is designed to provide precise speeds and tilt angles for a broad range of molecular and biological mixing applications. The user has the ability to adjust parameters to match the vessel size as well as the volume of liquid being mixed, yielding optimum results. The rocking speed and tilt angle are easily adjustable. At low speeds and minimal tilt angles, the BenchRocker 2D provides a gentle wave effect for non-foaming mixing for blot washing and gel work.