Learn More
Waters Corp WATERS CORPORATION OQ Qual Workbook, LAC/E Buffering, Module/Acquisition Client, Device Validation
Catalog No. 50466694 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The LAC/E Buffering OQ Qual Workbook is a recommended operational qualification procedure used to confirm LAC/E Buffering. This test verifies that data transfer between the acquisition device and the cloud is accomplished in a timely and accurate manner as part of device validation.
  • Verifies timely and accurate data transfer between the acquisition device and the cloud.
  • Ensures confirmation of LAC/E Buffering.
  • Supports device validation procedures.
  • Applicable after Edge Device software installation, upgrade, or removal.
Catalog No. 50-466-694 Supplier Waters Corp Supplier No. 715002971
The LAC/E Buffering OQ Qual Workbook is a recommended operational qualification procedure used to confirm LAC/E Buffering. This test verifies that data transfer between the acquisition device and the cloud is accomplished in a timely and accurate manner as part of device validation.
  • Verifies timely and accurate data transfer between the acquisition device and the cloud.
  • Ensures confirmation of LAC/E Buffering.
  • Supports device validation procedures.
  • Applicable after Edge Device software installation, upgrade, or removal.