Learn More
AdipoGen Lac-Phe Acetate
SDP
Supplier:  AdipoGen AGCR13545M050
Encompass_Preferred
Chemical. CAS 183241-73-8. Formula C12H15NO4 . C2H3O2. MW 237.3 . 59.0. Lac-Phe is an exercise-inducible metabolite, also called exerkine, that suppresses feeding and obesity. Lac-Phe is increased heavily with exercise in mammals and acts to reduce food intake and potentially decrease overall body weight and fat mass. In diet-induced obese mice, pharmacological administration of Lac-Phe led to decreased food intake, adiposity and body weight, as well as improved glucose homeostasis, while movement and energy expenditure were not affected. The biosynthesis of Lac-Phe from lactate and phenylalanine occurs in CNDP2 cells, including macrophages, monocytes and other immune and epithelial cells.
Catalog No. 50-256-2309