Learn More
Electron Microscopy Sciences EMS Lacey Carbon Film 50 µm holes on 400 mesh Cu
SDP
Supplier:  Electron Microscopy Sciences LC400CU
Encompass_Preferred
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Copper
Hole: 50 µm
Catalog No. 50-190-1074