Learn More
Ted Pella Inc Lacey Carbon, 200 mesh, TH, Copper, 50/pk
SDP
Catalog No. NC1934977
Encompass
Lacey Carbon Films with No Formvar: Holes are completely open.
Catalog No. NC1934977 Supplier Ted Pella Inc Supplier No. 01894F
Lacey Carbon Films with No Formvar: Holes are completely open.