Learn More
Ted Pella Inc Lacey Carbon, 300 mesh, Copper, approx. grid hole size: 63µm, 50/pk
SDP
Catalog No. NC1237959
Encompass
Lacey Carbon, 300 mesh, Copper, approx. grid hole size: 63µm. Holes are completely open.
Catalog No. NC1237959 Supplier Ted Pella Inc Supplier No. 01895F
Lacey Carbon, 300 mesh, Copper, approx. grid hole size: 63µm. Holes are completely open.