Learn More
Electron Microscopy Sciences EMS Lacey Carbon Film 100 µm holes on 300 mesh Cu
SDP
Catalog No. 501902742
Encompass_Preferred
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Copper
Hole: 100 µm
Catalog No. 50-190-2742 Supplier Electron Microscopy Sciences Supplier No. LC300CU100
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Copper
Hole: 100 µm