Learn More
Electron Microscopy Sciences Fischione Lacey Carbon on Copper Tomography Grid
SDP
Supplier:  Electron Microscopy Sciences 7435702
Encompass_Preferred
This 1.5mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90 deg.
Catalog No. 50-193-1247