Learn More
Electron Microscopy Sciences Fischione Lacey Carbon on Copper Tomography Grid
SDP
Catalog No. 501931246
Encompass_Preferred
This 1.5mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90 deg.
Catalog No. 50-193-1246 Supplier Electron Microscopy Sciences Supplier No. 7435701
This 1.5mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90 deg.