Learn More
Electron Microscopy Sciences EMS Lacey Carbon Film 4-6 nm thick on 200 mesh Nickel Grid
SDP
Catalog No. 501931306
Encompass_Preferred
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
Thickness: 4-6 nm
Catalog No. 50-193-1306 Supplier Electron Microscopy Sciences Supplier No. LC200NIUL
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
Thickness: 4-6 nm