Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaCrosse™ Economy Knee Boots

Catalog No. 19817609
Change view
Click to view available options
Size:
12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-817-609 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-817-609 Supplier LaCrosse™ Supplier No. 2400904312

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rugged boots provide great protection in muddy, grimy jobsites

  • Made of 100% waterproof, abrasion-resistant rubber
  • Double-layer, abrasion-resistant toe guard
  • Slip-resistant Diamond Bar™ outsole for excellent traction on varied surfaces
  • Steel shank
  • Boot height of 30.48cm (16 in.)

Compliance

Steel safety toe meets or exceeds ASTM standard F2413-11 M I/75 C/75 EH. Electrical hazard (EH) footwear is manufactured with non-conductive electrical shock resistant soles and heels

Specifications

Height (English) 16 in.
Material Rubber
Size 12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.