Learn More
LaCrosse™ Rainfair™ De-Icer™ Insulated Apparel
Catalog No. 19047620C
Change view
Click to view available options
Coverall; Black/Red; Large
Catalog No.
19-047-620C Coverall; Black/Red; Large
1 options
This item is not returnable. View return policy
Catalog No. 19-047-620C Supplier LaCrosse™ Supplier No. 18003800 LG
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Keep warm on the coldest days, without the extra weight of bulkier insulating materials
Insulated clothing features Thinsulate™ Lite Loft, exceptionally lightweight insulation that offers warmth without restricting movement.
  • Heavy-duty zippers and snaps ensure long-wearing durability
  • Roomy pockets provide ample storage for small tools and keys
  • Coveralls have an attached hood with drawcord, snap-over storm front, and snug knit cuffs to keep out the cold wind; red top, black bottom
Specifications
Color Black, Red