Learn More
LaCrosse™ Rainfair™ Power-Pro™ Brogue Dielectric Overshoes
Catalog No. 19099164F
Change view
Click to view available options
Size: 11
Catalog No.
19-099-164F Size: 11
1 options
This item is not returnable. View return policy
Catalog No. 19-099-164F Supplier LaCrosse™ Supplier No. 001441011
This item is not returnable. View return policy
Extra layer of rubber for increased electrical protection
Safety and comfort come together to create a brand of dielectric overshoes you can count on. Ideal for water and sewer applications involving utilities and power transmissions.
  • Constructed of durable ozone-resistant orange rubber with black rubber side reinforcements
  • Extra layer of rubber on the outside increases electrical hazard protection
  • Nylon/cotton net lining
  • Calendered rope roll oversized outsoles offer extra abrasion protection
  • Height: 14 in. (36cm)
Order Info
Made to fit size-to-size over work boots with oversized outsoles
Compliance
Exceeds ANSI™ Z41 PT91 EH standards at 20 kV for one minute