Learn More
Cayman Chemical LactacystIn 100UG
SDP
Catalog No. 502896595
Encompass_Preferred
A microbial metabolite isolated from Streptomyces that is widely used as a selective inhibitor of the 20S proteasome
Catalog No. 50-289-6595 Supplier Cayman Chemical Supplier No. 70980100 UG
A microbial metabolite isolated from Streptomyces that is widely used as a selective inhibitor of the 20S proteasome