Learn More
Cayman Chemical Lactacystin, 133343-34-7, 100 g
SDP
Supplier:  Cayman Chemical 70980100
Encompass_Preferred
Molecular Formula C15H24N2O7S, Purity ≥98%, Formula Weight 376.4; A microbial metabolite isolated from Streptomyces that is widely used as a selective inhibitor of the 20S proteasome.
Catalog No. NC0109985
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.