Learn More
AdipoGen Lactacystin, 133343-34-7, MFCD01076525, 200 ug
SDP
Catalog No. 50595736
Encompass_Preferred
MF: C15H24N2O7S, MW: 376.4, ≥98%, Appearance: White to off-white solid. Induces neuritogenesis, Apoptosis inducer, Anticancer compound, and Induces differentiation and inhibits cell cycle progression in several tumor cell lines.
Catalog No. 50-595-736 Supplier AdipoGen Supplier No. AGCN20104C200
MF: C15H24N2O7S, MW: 376.4, ≥98%, Appearance: White to off-white solid. Induces neuritogenesis, Apoptosis inducer, Anticancer compound, and Induces differentiation and inhibits cell cycle progression in several tumor cell lines.