Learn More
AdipoGen Lactacystin (500 µg)
SDP
Catalog No. 501489972
Encompass_Preferred
Lactacystin (500 µg), AdipoGen Life Sciences. Chemical. CAS: 133343-34-7. Formula: C15H24N2O7S. MW: 376.4. Isolated from Streptomyces lactacystinnaeus strain OM-6519. Induces neuritogenesis. Potent and selective irreversible proteasome inhibitor. Specifically inhibits 20S proteasome (MCP; multicatalytic complex). Blocks proteasome activity by targeting the catalytic beta-subunit. Apoptosis inducer. Anticancer compound. Upregulates HSP70 and HSP22. Induces differentiation and inhibits cell cycle progression in several tumor cell lines. Inhibits NF-kappaB activation by inhibiting IkappaB degradation. Cathepsin A inhibitor. beta-Secretase activity inhibitor. Anti-adipogenic. Autophagy inducer.
Catalog No. 50-148-9972 Supplier AdipoGen Supplier No. AGCN20104C500
Lactacystin (500 µg), AdipoGen Life Sciences. Chemical. CAS: 133343-34-7. Formula: C15H24N2O7S. MW: 376.4. Isolated from Streptomyces lactacystinnaeus strain OM-6519. Induces neuritogenesis. Potent and selective irreversible proteasome inhibitor. Specifically inhibits 20S proteasome (MCP; multicatalytic complex). Blocks proteasome activity by targeting the catalytic beta-subunit. Apoptosis inducer. Anticancer compound. Upregulates HSP70 and HSP22. Induces differentiation and inhibits cell cycle progression in several tumor cell lines. Inhibits NF-kappaB activation by inhibiting IkappaB degradation. Cathepsin A inhibitor. beta-Secretase activity inhibitor. Anti-adipogenic. Autophagy inducer.