Learn More
Abcam Lactacystin, proteasome inhibitor, 200UG
Catalog No. 502583359
Encompass_Preferred
MW 376.4 Da, Purity >95%. Potent, irreversible, cell-permeable, selective 20S proteasome inhibitor (IC₅₀ = 70 nM). Inhibits cell-cycle progression and induces neurite outgrowth in vitro. Anti-bacterial and antitumor agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3359 Supplier Abcam Supplier No. AB141411200UG
MW 376.4 Da, Purity >95%. Potent, irreversible, cell-permeable, selective 20S proteasome inhibitor (IC₅₀ = 70 nM). Inhibits cell-cycle progression and induces neurite outgrowth in vitro. Anti-bacterial and antitumor agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement