Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactalbumin hydrolysate

Catalog No. AAJ6321536
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6321536 500 g
AAJ63215A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6321536 Supplier Thermo Scientific Chemicals Supplier No. J6321536
Add to Cart

A solubilized enzymatic digest of lactalbumin | CAS: 68458-87-7

Lactalbumin hydrolysate is used as a highly soluble source of amino acids and peptides. It is especially useful in fermentations, laboratory media and in tissue culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lactalbumin hydrolysate is used as a highly soluble source of amino acids and peptides. It is especially useful in fermentations, laboratory media and in tissue culture.

Solubility
Soluble in water (130 mg/ml at 25°C).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68458-87-7
MDL Number MFCD00147378
Synonym Enzymatic Digest of Lactalbumin

Specifications

Color Tan
Quantity 500 g
Solubility Information Soluble in water (130mg/ml at 25°C).
Physical Form Powder
Chemical Name or Material Lactalbumin hydrolysate
EINECSNumber 270-615-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.