Learn More
Medchemexpress LLC HY-138247 5mg Medchemexpress, β-Lactamase-IN-2 CAS:2114651-20-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1382475MG
Encompass_Preferred
Medchemexpress, HY-138247 5mg β-Lactamase-IN-2 CAS:2114651-20-4 β-Lactamase-IN-2 is a beta-lactamase inhibitor, extracted from patent WO 2019075084 A1, compound 1. β-Lactamase-IN-2 has anti-microbial and anti-bacterial effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1520