Learn More
Dojindo Molecular Technologies Inc Lactate Assay Kit -WST (50 TESTS), Quantification of L-lactate levels and it is used to monitor changes intracellular metabolic pathways of cancer reaserch.
SDP
Catalog No. 501903722
Encompass_Preferred
Lactate is a metabolite of glycolysis that is one of the main metabolic pathways in cells, and is known to be a biomarker for muscular fatigue and hyperlactacidemia. It also serves as a marker for monitoring the changes of intracellular metabolic pathways. In addition, recent metabolomic study suggests that lactate contributes as a major carbon source in the TCA cycle of tissues and cancer cells. Lactate Assay Kit-WST enables quantitation of lactate produced by glycolysis. This kit has been optimized to quantitate lactate in cell culture supernatant by measuring the absorption derived from a colorimetric reaction of WST. This kit is formated for 96-well microplate assays with a detection sensitivity limit of 0.02 mmol/l lactate. Dojindo has various quantification kits of Intracellular Metabolites. (https://www.dojindo.com/quantification-of-intracellular-metabolites/)
Catalog No. 50-190-3722 Supplier Dojindo Molecular Technologies Inc Supplier No. L25610
Lactate is a metabolite of glycolysis that is one of the main metabolic pathways in cells, and is known to be a biomarker for muscular fatigue and hyperlactacidemia. It also serves as a marker for monitoring the changes of intracellular metabolic pathways. In addition, recent metabolomic study suggests that lactate contributes as a major carbon source in the TCA cycle of tissues and cancer cells. Lactate Assay Kit-WST enables quantitation of lactate produced by glycolysis. This kit has been optimized to quantitate lactate in cell culture supernatant by measuring the absorption derived from a colorimetric reaction of WST. This kit is formated for 96-well microplate assays with a detection sensitivity limit of 0.02 mmol/l lactate. Dojindo has various quantification kits of Intracellular Metabolites. (https://www.dojindo.com/quantification-of-intracellular-metabolites/)