Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lactate-d3 sodium | 1219802-24-0 | MFCD29050090 | 98.0% | 115.08 g/mol | C3H2D3NaO3 | 10 MG
SDP

Catalog No. 5000297312
Encompass_Preferred

Lactate-d3 sodium is the deuterium-labeled sodium salt of DL-lactic acid supplied as a 60% w/w aqueous solution for use as an isotopically labeled reagent in analytical and metabolic studies.

  • Supplied as a 60% w/w aqueous solution suitable for stock and dilution.
  • Deuterium-labeled (d3) sodium salt for isotopic tracing and mass spectrometry applications.
  • High purity: 98.0%.
  • Molecular weight approximately 115.08 g/mol and formula C3H2D3NaO3.
  • Provided in a small 10 mg quantity for analytical use and method development.

Catalog No. 50-002-97312 Supplier Medchemexpress LLC Supplier No. HYB2227BS110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lactate-d3 sodium is the deuterium-labeled sodium salt of DL-lactic acid supplied as a 60% w/w aqueous solution for use as an isotopically labeled reagent in analytical and metabolic studies.

  • Supplied as a 60% w/w aqueous solution suitable for stock and dilution.
  • Deuterium-labeled (d3) sodium salt for isotopic tracing and mass spectrometry applications.
  • High purity: 98.0%.
  • Molecular weight approximately 115.08 g/mol and formula C3H2D3NaO3.
  • Provided in a small 10 mg quantity for analytical use and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.