Learn More
Medchemexpress LLC Lactate-d4 sodium | 344299-52-1 | 99.0% | 5 MG
SDP
Catalog No. 5000409687
Encompass_Preferred
Lactic acid-d4 sodium (60% in water) is a deuterium-labeled form of lactic acid sodium. The unlabeled compound is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator that induces lysine residue lactylation. It serves as a glycolysis end-product, bridging glycolysis and oxidative phosphorylation. It is also recognized as an oncometabolite with an immune-protective role in anti-tumor immunity and possesses antimicrobial activity, making it suitable as a food preservative.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-09687 Supplier Medchemexpress LLC Supplier No. HYB2227BS5MG
Lactic acid-d4 sodium (60% in water) is a deuterium-labeled form of lactic acid sodium. The unlabeled compound is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator that induces lysine residue lactylation. It serves as a glycolysis end-product, bridging glycolysis and oxidative phosphorylation. It is also recognized as an oncometabolite with an immune-protective role in anti-tumor immunity and possesses antimicrobial activity, making it suitable as a food preservative.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.