Learn More
Cusabio Technology LLC lactate dehydrogenase D
Catalog No. 5000048937
Encompass_Preferred
The Human D-Lactate Dehydrogenase (D-LDH) ELISA Kit is designed for the quantitative detection of LDHD in serum plasma and tissue homogenates using a sandwich assay method This kit detects LDHD with a sensitivity of 7 8 mIU/mL and provides a broad detection range from 31 25 mIU/mL to 2000 mIU/mL The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable and precise quantification of D-LDH levels in various biological samples Suitable for research and diagnostic applications this kit offers a standardized approach to analyze lactate dehydrogenase activity with high specificity and reproducibility
Catalog No. 50-000-48937 Supplier Cusabio Technology LLC Supplier No. CSBE11720H
The Human D-Lactate Dehydrogenase (D-LDH) ELISA Kit is designed for the quantitative detection of LDHD in serum plasma and tissue homogenates using a sandwich assay method This kit detects LDHD with a sensitivity of 7 8 mIU/mL and provides a broad detection range from 31 25 mIU/mL to 2000 mIU/mL The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable and precise quantification of D-LDH levels in various biological samples Suitable for research and diagnostic applications this kit offers a standardized approach to analyze lactate dehydrogenase activity with high specificity and reproducibility