Learn More
Sigma Aldrich Fine Chemicals Biosciences Lactate Dehydrogenase Activity Assay Kit
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK0661KT
Encompass
Lactate Dehydrogenase Activity Assay Kit has been used for measuring the activity of lactate dehydrogenase (LDH) in the cell culture medium.
Catalog No. NC1812150