Learn More
Apexbio Technology LLC Lactate-Fluoro-Assay-Kit
SDP
Catalog No. 5000567391
Encompass_Preferred
Lactate Fluorometric Assay Kit is a vital tool in biomedical research originating from the need to accurately measure lactate levels in biological samples The kit operates by targeting the lactate-to-pyruvate pathway utilizing a fluorometric method to detect changes in fluorescence indicating lactate concentration Its sensitivity often reaches the nanomolar to low micromolar range making it highly effective for in vitro assays Commonly applied in cell models and animal studies this kit is particularly valuable in drug screening processes where it helps assess metabolic function and cellular respiration Experimental concentrations typically depend on the specific research design allowing for versatile application across various studies focused on metabolic disorders cancer research and physiological monitoring ensuring researchers have precise and reliable data
Catalog No. 50-005-67391 Supplier Apexbio Technology LLC Supplier No. K2140100
Add to Cart
Add to Cart
Lactate Fluorometric Assay Kit is a vital tool in biomedical research originating from the need to accurately measure lactate levels in biological samples The kit operates by targeting the lactate-to-pyruvate pathway utilizing a fluorometric method to detect changes in fluorescence indicating lactate concentration Its sensitivity often reaches the nanomolar to low micromolar range making it highly effective for in vitro assays Commonly applied in cell models and animal studies this kit is particularly valuable in drug screening processes where it helps assess metabolic function and cellular respiration Experimental concentrations typically depend on the specific research design allowing for versatile application across various studies focused on metabolic disorders cancer research and physiological monitoring ensuring researchers have precise and reliable data