Learn More
AG Scientific Inc Lactate Oxidase, 1 KU
Catalog No. 502236951
Encompass_Preferred
Lactate oxidase is a flavin mononucleotide-dependent alpha hydroxyl acid oxidizing enzyme. Lactate oxidase is useful for enzymatic determination of lactic acid. It uses molecular oxygen to catalyze the oxidation of L-lactate.Lactate oxidase enzymes belong to the family of oxidoreductase which act on single donors with O2 as oxidants. The enzymes often come from Aerococcus viridian and appear in viruses and cellular organisms.CAS Number: 9028-72-2Molecular Weight: 80 kDa (gel filtration)Storage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6951 Supplier AG Scientific Inc Supplier No. L11751KU
Lactate oxidase is a flavin mononucleotide-dependent alpha hydroxyl acid oxidizing enzyme. Lactate oxidase is useful for enzymatic determination of lactic acid. It uses molecular oxygen to catalyze the oxidation of L-lactate.Lactate oxidase enzymes belong to the family of oxidoreductase which act on single donors with O2 as oxidants. The enzymes often come from Aerococcus viridian and appear in viruses and cellular organisms.CAS Number: 9028-72-2Molecular Weight: 80 kDa (gel filtration)Storage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.