Learn More
Sports Resource Group Inc LACTATE PLUS TEST STRIP 25/PK
Catalog No. NC0071872
Encompass
Lactate Plus Test Strips, vial of 25. Compatible with the Lactate Plus Analyzer. No coding or refrigeration necessary. Made in USA.
Catalog No. NC0071872 Supplier Sports Resource Group Inc Supplier No. 40813
Lactate Plus Test Strips, vial of 25. Compatible with the Lactate Plus Analyzer. No coding or refrigeration necessary. Made in USA.