Learn More
Nova Biomedical Corporation Nova Biomedical Lactate Sensor
Catalog No. NC9846021
Encompass
LACTATE ELECTRODE, BIOPROFILE LACTATE ELECTRODE, BIOPROFILELactate Electrode For BioProfile 300A / 300B / 400 and Profile 100
Catalog No. NC9846021 Supplier Nova Biomedical Corporation Supplier No. 24449
LACTATE ELECTRODE, BIOPROFILE LACTATE ELECTRODE, BIOPROFILELactate Electrode For BioProfile 300A / 300B / 400 and Profile 100