Learn More
McKesson General Medical Lactated Ringer's Solution, Sterile IV, Electrolyte Balance, 1000 mL per Unit
Catalog No. NC3235560
Encompass
Lactated Ringer's Solution is an isotonic intravenous solution supplied in case of 14 x 1000 mL units, designed for fluid and electrolyte replenishment. It closely matches extracellular fluid composition, ideal for addressing fluid losses and electrolyte imbalances. This sterile, nonpyrogenic solution is free from latex, PVC, and DEHP, ensuring safety for patients. Its formulation includes essential electrolytes and lactate, offering a metabolic alkalinizing effect, distinguishing it from other IV solutions.
Catalog No. NC3235560 Supplier McKesson General Medical Supplier No. 515492
Lactated Ringer's Solution is an isotonic intravenous solution supplied in case of 14 x 1000 mL units, designed for fluid and electrolyte replenishment. It closely matches extracellular fluid composition, ideal for addressing fluid losses and electrolyte imbalances. This sterile, nonpyrogenic solution is free from latex, PVC, and DEHP, ensuring safety for patients. Its formulation includes essential electrolytes and lactate, offering a metabolic alkalinizing effect, distinguishing it from other IV solutions.