Learn More
Aviva Systems Biology Corp LACTB Antibody - middle region (ARP52430_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52430_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LACTB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LACTB. Alternate names: FLJ14902, G24, MRPL56ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60740