Learn More
Aviva Systems Biology Corp LACTB2 Antibody - middle region (ARP56810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LACTB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LACTB2. Alternate names: CGI-83ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61997