Learn More
Apexbio Technology LLC Lactic acid(Synonyms: 2-Hydroxypropanoic acid, α-Hydroxypropionic acid, Milk acid, DL-Lactic acid), 100mg, CAS: 50-21-5.
SDP
Catalog No. 5000060874
Encompass_Preferred
Lactic acid (CAS 50-21-5) is an organic compound formed naturally as an intermediate during anaerobic glycolysis where sugars are metabolized through fermentation Biologically lactic acid reduces local pH environments thus inhibiting pathogenic microbial growth Due to this acidifying action it is utilized in biomedical contexts including formulations aimed at preventing gastrointestinal microbial fermentation Additionally lactic acid served as one of the active ingredients in Phexxi a non-hormonal contraceptive approved by the FDA in May 2020 wherein it maintains vaginal acidity to exert contraceptive effects
Catalog No. 50-000-60874 Supplier Apexbio Technology LLC Supplier No. B7893100
Lactic acid (CAS 50-21-5) is an organic compound formed naturally as an intermediate during anaerobic glycolysis where sugars are metabolized through fermentation Biologically lactic acid reduces local pH environments thus inhibiting pathogenic microbial growth Due to this acidifying action it is utilized in biomedical contexts including formulations aimed at preventing gastrointestinal microbial fermentation Additionally lactic acid served as one of the active ingredients in Phexxi a non-hormonal contraceptive approved by the FDA in May 2020 wherein it maintains vaginal acidity to exert contraceptive effects