Learn More
Medchemexpress LLC Lactic acid | 50-21-5 | 85.1% | 100 MG
SDP
Catalog No. 5000410639
Encompass_Preferred
Lactic acid (DL-Lactic acid) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator that induces lactylation of lysine residues. It is a glycolysis end-product that links glycolysis and oxidative phosphorylation. This product is considered an oncometabolite, plays an immune protective role in anti-tumor immunity, and also possesses antimicrobial activity.
  • Activates HCAR1.
  • Induces lysine residues lactylation.
  • Serves as a glycolysis end-product.
  • Bridges glycolysis and oxidative phosphorylation.
  • Demonstrates immune protective role in anti-tumor immunity.
  • Exhibits antimicrobial activity.
  • Can be used as a food preservative.
Catalog No. 50-004-10639 Supplier Medchemexpress LLC Supplier No. HYB2227100MG
Lactic acid (DL-Lactic acid) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator that induces lactylation of lysine residues. It is a glycolysis end-product that links glycolysis and oxidative phosphorylation. This product is considered an oncometabolite, plays an immune protective role in anti-tumor immunity, and also possesses antimicrobial activity.
  • Activates HCAR1.
  • Induces lysine residues lactylation.
  • Serves as a glycolysis end-product.
  • Bridges glycolysis and oxidative phosphorylation.
  • Demonstrates immune protective role in anti-tumor immunity.
  • Exhibits antimicrobial activity.
  • Can be used as a food preservative.