Learn More
Medchemexpress LLC Lactic acid | 50-21-5 | 85.1% | 500 MG
SDP
Catalog No. 5000410553
Encompass_Preferred
Lactic acid (DL-Lactic acid) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. Lactic acid is an oncometabolite and has an immune protective role of lactate in anti-tumor immunity. It also possesses antimicrobial activity, which allows its use as a food preservative.
  • Hydroxycarboxylic acid receptor 1 (HCAR1) activator
  • Epigenetic modulator inducing lysine residues lactylation
  • Glycolysis end-product
  • Oncometabolite
  • Immune protective role of lactate in anti-tumor immunity
  • Antimicrobial activity (can be used as a food preservative)
Catalog No. 50-004-10553 Supplier Medchemexpress LLC Supplier No. HYB2227500MG
Lactic acid (DL-Lactic acid) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. Lactic acid is an oncometabolite and has an immune protective role of lactate in anti-tumor immunity. It also possesses antimicrobial activity, which allows its use as a food preservative.
  • Hydroxycarboxylic acid receptor 1 (HCAR1) activator
  • Epigenetic modulator inducing lysine residues lactylation
  • Glycolysis end-product
  • Oncometabolite
  • Immune protective role of lactate in anti-tumor immunity
  • Antimicrobial activity (can be used as a food preservative)