Learn More
Alkali Scientific Lactic Acid 88% Solution, 1 L
SDP
Catalog No. 5000659924
Encompass_Preferred
Lactic Acid 88% Solution is a high-concentration reagent used for pH adjustment and buffer preparation in laboratory applications. This 1-liter bottle is ideal for applications requiring precise pH control, particularly in studies related to acid-base reactions and metabolic processes. Lactic acid is commonly used in chemical synthesis, as well as in the preparation of buffers and solutions for microbiological and biochemical studies. Its effectiveness in adjusting pH to the required levels makes it an essential tool in various laboratory procedures, including protein purification, cell culture, and enzymatic assays where specific acidic conditions are necessary for optimal reaction conditions.
Catalog No. 50-006-59924 Supplier Alkali Scientific Supplier No. LAC001CP
Lactic Acid 88% Solution is a high-concentration reagent used for pH adjustment and buffer preparation in laboratory applications. This 1-liter bottle is ideal for applications requiring precise pH control, particularly in studies related to acid-base reactions and metabolic processes. Lactic acid is commonly used in chemical synthesis, as well as in the preparation of buffers and solutions for microbiological and biochemical studies. Its effectiveness in adjusting pH to the required levels makes it an essential tool in various laboratory procedures, including protein purification, cell culture, and enzymatic assays where specific acidic conditions are necessary for optimal reaction conditions.