Learn More
Alkali Scientific Lactic Acid 88% Solution, 500 Milliliters
SDP
Catalog No. 5000659743
Encompass_Preferred
Lactic acid 88% solution is a highly concentrated reagent used in pH adjustment applications. This 500mL solution is commonly employed in laboratory processes that require precise acidification of solutions to maintain optimal pH levels for biological or chemical reactions. Lactic acid is widely used in fermentation processes, food preservation, and in the production of lactic acid-based polymers. Its ability to lower pH without adding excess ionic strength makes it a suitable choice for sensitive biological applications, such as cell culture and enzymatic assays. This solution is often used in research laboratories, industrial applications, and bioprocessing environments.
Catalog No. 50-006-59743 Supplier Alkali Scientific Supplier No. LAC003CP
Lactic acid 88% solution is a highly concentrated reagent used in pH adjustment applications. This 500mL solution is commonly employed in laboratory processes that require precise acidification of solutions to maintain optimal pH levels for biological or chemical reactions. Lactic acid is widely used in fermentation processes, food preservation, and in the production of lactic acid-based polymers. Its ability to lower pH without adding excess ionic strength makes it a suitable choice for sensitive biological applications, such as cell culture and enzymatic assays. This solution is often used in research laboratories, industrial applications, and bioprocessing environments.