Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lactic acid, Biotin labeled | 98.0% | 100 MG
SDP

Catalog No. 5000410956
Encompass_Preferred

Lactic acid, Biotin labeled is a biotin-labeled form of lactic acid, specifically DL-Lactic acid, Biotin labeled. It is designed to be conjugated to streptavidin for purification processes. This product is intended for research use only and is suitable for various biochemical assay applications.

  • Purity of 98.0%
  • Molecular weight of 358.46
  • Molecular formula: C15H26N4O4S
  • Appears as an off-white to light yellow solid

Catalog No. 50-004-10956 Supplier Medchemexpress LLC Supplier No. HYB2227F100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lactic acid, Biotin labeled is a biotin-labeled form of lactic acid, specifically DL-Lactic acid, Biotin labeled. It is designed to be conjugated to streptavidin for purification processes. This product is intended for research use only and is suitable for various biochemical assay applications.

  • Purity of 98.0%
  • Molecular weight of 358.46
  • Molecular formula: C15H26N4O4S
  • Appears as an off-white to light yellow solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.