Learn More
Medchemexpress LLC Lactic acid, Biotin labeled | 98.0% | 25 MG
SDP
Catalog No. 5000412621
Encompass_Preferred
Lactic acid, Biotin labeled, also known as DL-Lactic acid, Biotin labeled, is a form of lactic acid that has been labeled with biotin. This biotinylated lactic acid can be utilized for conjugation to streptavidin, facilitating various purification processes.
  • Biotin-labeled lactic acid for research applications.
  • Can be conjugated to streptavidin for purification.
  • Suitable for laboratory use.
Catalog No. 50-004-12621 Supplier Medchemexpress LLC Supplier No. 25MGHYB2227F
Lactic acid, Biotin labeled, also known as DL-Lactic acid, Biotin labeled, is a form of lactic acid that has been labeled with biotin. This biotinylated lactic acid can be utilized for conjugation to streptavidin, facilitating various purification processes.
  • Biotin-labeled lactic acid for research applications.
  • Can be conjugated to streptavidin for purification.
  • Suitable for laboratory use.