Learn More
Medchemexpress LLC Lactic acid, biotin labeled | 98.0% | 358.46 | C15H26N4O4S | 10 MG
SDP
Catalog No. 5000297027
Encompass_Preferred
Lactic acid, biotin labeled is a biotin-conjugated form of DL-lactic acid intended for research use. The biotin tag enables affinity capture and detection with streptavidin-based reagents. Supplied as a dry powder in milligram-scale quantities, it is suitable for affinity purification and detection workflows and is typically used in biochemical assays requiring a biotinylated small molecule.
Catalog No. 50-002-97027 Supplier Medchemexpress LLC Supplier No. HYB2227F10MG
Lactic acid, biotin labeled is a biotin-conjugated form of DL-lactic acid intended for research use. The biotin tag enables affinity capture and detection with streptavidin-based reagents. Supplied as a dry powder in milligram-scale quantities, it is suitable for affinity purification and detection workflows and is typically used in biochemical assays requiring a biotinylated small molecule.