Learn More
Medchemexpress LLC Lactic acid, Biotin labeled | 98.0% | 50 MG
SDP
Catalog No. 5000410957
Encompass_Preferred
Lactic acid, Biotin labeled is a biotin-labeled form of lactic acid. It can be conjugated to streptavidin for purification processes in biochemical assays.
  • Biotin labeled lactic acid
  • Conjugates to streptavidin
  • Supports purification applications
  • Suitable for biochemical assays
Catalog No. 50-004-10957 Supplier Medchemexpress LLC Supplier No. HYB2227F50MG
Lactic acid, Biotin labeled is a biotin-labeled form of lactic acid. It can be conjugated to streptavidin for purification processes in biochemical assays.
  • Biotin labeled lactic acid
  • Conjugates to streptavidin
  • Supports purification applications
  • Suitable for biochemical assays