Learn More
Medchemexpress LLC Lactic acid-d3 sodium | 1219802-24-0 | 98% | 5 MG
SDP
Catalog No. 5000409688
Encompass_Preferred
Lactic acid-d3 sodium is the deuterium labeled form of Lactic acid sodium, an HCAR1 activator and epigenetic modulator. It plays a role as a glycolysis end-product, bridging glycolysis and oxidative phosphorylation. This compound is also recognized as an oncometabolite with an immune protective role in anti-tumor immunity and possesses antimicrobial activity, making it suitable as a food preservative.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09688 Supplier Medchemexpress LLC Supplier No. HYB2227BS15MG
Lactic acid-d3 sodium is the deuterium labeled form of Lactic acid sodium, an HCAR1 activator and epigenetic modulator. It plays a role as a glycolysis end-product, bridging glycolysis and oxidative phosphorylation. This compound is also recognized as an oncometabolite with an immune protective role in anti-tumor immunity and possesses antimicrobial activity, making it suitable as a food preservative.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.