Learn More
Medchemexpress LLC Lactic acid (DL-lactic acid) | 50-21-5 | 85.1% | 5 G
SDP
Catalog No. 5000410543
Encompass_Preferred
Lactic acid (DL-Lactic acid) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging glycolysis and oxidative phosphorylation. Lactic acid is an oncometabolite and has an immune protective role in anti-tumor immunity. It also possesses antimicrobial activity and can be used as a food preservative.
  • Activates hydroxycarboxylic acid receptor 1 (HCAR1).
  • Induces lysine residues lactylation.
  • Glycolysis end-product.
  • Bridging glycolysis and oxidative phosphorylation.
  • Oncometabolite.
  • Immune protective role in anti-tumor immunity.
  • Possesses antimicrobial activity.
  • Can be used as a food preservative.
Catalog No. 50-004-10543 Supplier Medchemexpress LLC Supplier No. HYB22275G
Lactic acid (DL-Lactic acid) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging glycolysis and oxidative phosphorylation. Lactic acid is an oncometabolite and has an immune protective role in anti-tumor immunity. It also possesses antimicrobial activity and can be used as a food preservative.
  • Activates hydroxycarboxylic acid receptor 1 (HCAR1).
  • Induces lysine residues lactylation.
  • Glycolysis end-product.
  • Bridging glycolysis and oxidative phosphorylation.
  • Oncometabolite.
  • Immune protective role in anti-tumor immunity.
  • Possesses antimicrobial activity.
  • Can be used as a food preservative.